Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900663

Sigma-Aldrich

Cyclooctanone

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14(=O)
CAS Number:
Molecular Weight:
126.20
Beilstein/REAXYS Number:
1280738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

195-197 °C (lit.)

mp

32-41 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCC1

InChI

1S/C8H14O/c9-8-6-4-2-1-3-5-7-8/h1-7H2

InChI key

IIRFCWANHMSDCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yamada et al.
Chemical & pharmaceutical bulletin, 45(12), 2113-2115 (1998-01-20)
Stereoselective syntheses of omega-(alpha-bromoketo) octanals and nonanal with oxygenated functions and formation of the corresponding eight-membered carbocyclic aldols by subsequent samarium(II)-mediated cyclization are demonstrated. Cyclooctenones deoxygenated at the C2 or C10 position in the taxane framework are prepared by dehydration
Lei Jiao et al.
Journal of the American Chemical Society, 130(23), 7178-7179 (2008-05-16)
Vinylcyclopropane (VCP) has been well applied as a five-carbon component, rather than a three-carbon component, in transition-metal catalyzed cycloadditions. Here we demonstrate a Rh(I)-catalyzed [3 + 2] reaction of trans-VCP-enes, where VCP acts as a three-carbon synthon to furnish five-membered
Korany A Ali et al.
Archiv der Pharmazie, 345(3), 231-239 (2011-11-03)
The versatile synthon (E)-2-((dimethyl amino)methylene)cyclooctanone (2) was used as a key intermediate for the synthesis of cyclooctanones and cyclooctane-based heterocycles with pyrazole, isoxazole, pyrimidine, pyrazolopyrimidine, triazolopyrimidine and imidazopyrimidine derivatives via its reactions with several nitrogen nucleophiles. The newly synthesized compounds
Wei Zou et al.
Carbohydrate research, 339(15), 2475-2485 (2004-10-13)
Whereas C-2- and 4-ulopyranosyl compounds (C-2- and C-4-ulosides) can be converted to cyclopentenones under base conditions through beta-elimination and ring contraction, base-initiated beta-elimination of C-glycosyl 2'-aldehydes and 2'-ketones results in the formation of acyclic alpha,beta-unsaturated aldehydes or ketones. By combining
K Yamada et al.
Chemical & pharmaceutical bulletin, 45(12), 1898-1905 (1998-01-20)
Construction of the AB-ring system of the taxane framework via an A-ring annulation strategy was demonstrated by base-mediated intramolecular aldol reaction of (Z)-2,2-dimethyl-3-(1-methyl-2-oxopropylidene)cyclooctanone, affording the title compound, 1-hydroxy-8,11,11-trimethylbicyclo[5.3.1]undec-7-en-9-one. A cyclization precursor, the tetra-substituted (Z)-alkene, was prepared from the corresponding cyclooctanone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service