Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900661

Sigma-Aldrich

4-Methyl-o-phenylenediamine

Vetec, reagent grade, 97%

Synonym(s):

3,4-Toluenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein/REAXYS Number:
507965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

bp

155-156 °C/18 mmHg (lit.)

mp

87-89 °C (lit.)

SMILES string

Cc1ccc(N)c(N)c1

InChI

1S/C7H10N2/c1-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3

InChI key

DGRGLKZMKWPMOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Becci et al.
Toxicology and applied pharmacology, 71(3), 323-329 (1983-12-01)
o-Toluenediamine in corn oil was administered po to Sprague-Dawley rats at dosages of 10, 30, 100, or 300 mg/kg body wt/day during Days 6 through 15 of gestation. All animals were killed on Day 20 of gestation. A similar study
Effect of 4 toluene diamine isomers on murine testicular DNA synthesis.
E J Greene et al.
Mutation research, 91(1), 75-79 (1981-01-01)
T A Marks et al.
Teratology, 24(3), 253-265 (1981-12-01)
Pregnant outbred albino (CD-1) mice were given 2-nitro-p-phenylenediamine (2NPPD; 32-256 mg/kg/day), 4-nitro-o-phenylenediamine (4NOPD; 16-1024 mg/kg/day) or 2,5-toluenediamine sulfate (2,5TDS; 16-64 mg/kg/day) by subcutaneous injection on Days 6-15 of gestation. The mice were killed on Day 18, the general health and
Christina L Burnett et al.
International journal of toxicology, 29(3 Suppl), 61S-83S (2010-05-22)
Toluene-2,5-diamine, toluene-2,5-diamine sulfate, and toluene-3,4-diamine are diaminotoluenes used as colorants in permanent hair dyes and tints. Toluene-2,5-diamine is used in 79 products at concentrations up to 3%; toluene-2,5-diamine sulfate is used in 168 products at concentrations up to 4%. Toluene-3,4-diamine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service