Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900659

Sigma-Aldrich

Mesitaldehyde

Vetec, reagent grade, 98%

Synonym(s):

2,4,6-Trimethylbenzaldehyde, 2-Formyl-1,3,5-trimethylbenzene, 2-Formylmesitylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CHO
CAS Number:
Molecular Weight:
148.20
Beilstein/REAXYS Number:
1364114
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.553 (lit.)

bp

237 °C (lit.)

mp

10-12 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(C=O)c(C)c1

InChI

1S/C10H12O/c1-7-4-8(2)10(6-11)9(3)5-7/h4-6H,1-3H3

InChI key

HIKRJHFHGKZKRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service