Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900656

Sigma-Aldrich

Trimethyl-tetradecylammonium chloride

Vetec, reagent grade, 97%

Synonym(s):

Tetradecyltrimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Cl)(CH3)3
CAS Number:
Molecular Weight:
291.94
Beilstein/REAXYS Number:
3916572
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

~250 °C (dec.)

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CEYYIKYYFSTQRU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Zhang et al.
The journal of physical chemistry. B, 114(41), 13128-13135 (2010-09-28)
Two fatty acids, perfluorononanoic acid (C(8)F(17)COOH) and nonanoic acid (C(8)H(17)COOH), were mixed with a cationic hydrocarbon surfactant, tetradecyltrimethylammonium hydroxide (TTAOH), in aqueous solutions for comparative investigation. Phase behaviors of the two systems are quite different because of the special properties
Mohd Sajid Ali et al.
Journal of colloid and interface science, 352(2), 436-443 (2010-09-25)
Unfolding of rabbit serum albumin (RSA) by cationic surfactants cetyltrimethylammonium bromide (CTAB) and tetradecyltrimethylammonium bromide (TTAB) was studied by exploiting surface tensiometry, small-angle neutron scattering (SANS), intrinsic fluorescence, resonance Rayleigh scattering (RRS) (also referred as turbidity at 350/350), and circular
Suruchi Mahajan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17238-17246 (2012-12-12)
Keeping in view the use of surfactants in drug delivery, the interactions of surface active ionic liquids, such as 1-tetradecyl-3-methylimidazolium bromide (C(14)mimBr), with drugs, viz., dopamine hydrochloride (DH) and acetylcholine chloride (AC), have been studied, and the results are further
Dulan B Gunasekara et al.
Electrophoresis, 32(8), 832-837 (2011-03-26)
The combination of microchip electrophoresis with amperometric detection leads to a number of analytical challenges that are associated with isolating the detector from the high voltages used for the separation. While methods such as end-channel alignment and the use of
Yuri Borodko et al.
Langmuir : the ACS journal of surfaces and colloids, 25(12), 6665-6671 (2009-04-29)
The vibrational spectra of platinum nanoparticles (12 nm) capped with tetradecyltrimethylammonium bromide, C(14)TAB, were investigated by Fourier transform infrared (FTIR) spectroscopy. We have shown that the thermal decay of Pt-C(14)TAB nanoparticles in N(2), H(2), and O(2) atmospheres leads to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service