Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900653

Sigma-Aldrich

2,4,5-Trimethoxybenzaldehyde

Vetec, reagent grade, 98%

Synonym(s):

Asaronaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CHO
CAS Number:
Molecular Weight:
196.20
Beilstein/REAXYS Number:
1951403
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

112-114 °C (lit.)

SMILES string

[H]C(=O)c1cc(OC)c(OC)cc1OC

InChI

1S/C10H12O4/c1-12-8-5-10(14-3)9(13-2)4-7(8)6-11/h4-6H,1-3H3

InChI key

IAJBQAYHSQIQRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C-C Chen et al.
Letters in applied microbiology, 44(4), 387-392 (2007-04-03)
To investigate the active ingredient in fruiting bodies and to produce it with cultured mycelium in Antrodia camphorata (BCRC 35398). The volatile components from the fruiting bodies, the liquid cultured broth of A. camphorata and Cinnamomum kanehirae wood were separately
Man-Ru Wu et al.
Journal of agricultural and food chemistry, 60(29), 7262-7269 (2012-07-04)
Obesity is a global health problem. Because of the high costs and side effects of obesity-treatment drugs, the potential of natural products as alternatives for treating obesity is under exploration. 2,4,5-Trimethoxybenzaldehyde (2,4,5-TMBA) present in plant roots, seeds, and leaves was
S J Kim et al.
Microbial pathogenesis, 21(1), 65-70 (1996-07-01)
A 30 kDa (heated 24 kDa) hemin-binding protein whose expression is both hemin and iron regulated was identified and purified in Porphyromonas gingivalis 381. A strong hemin-binding function was found by LDS-PAGE and TMBZ staining when cells were grown under
Alexandro Silva Nunes et al.
Experimental parasitology, 135(4), 661-668 (2013-10-22)
Meloidogyne exigua is a parasitic nematode of plants that causes great losses to coffee farmers. In an effort to develop parasitic controls, 154 chalcones were synthesized and screened for activity against this nematode. The best results were obtained with (2E)-1-(4'-nitrophenyl)-3-(2,4,5-trimethoxyphenyl)prop-2-en-1-one
J J Cypher et al.
Biochemical genetics, 20(3-4), 315-332 (1982-04-01)
The substrate specificities of aldehyde and pyridoxal oxidases in Drosophila melanogaster have been determined with a variety of aliphatic and aromatic aldehydes. This analysis has led to the discovery that 2,4,5-trimethoxybenzaldehyde is a specific substrate for pyridoxal oxidase, as based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service