Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900651

Sigma-Aldrich

4-(Benzyloxy)benzaldehyde

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CHO
CAS Number:
Molecular Weight:
212.24
Beilstein/REAXYS Number:
1242385
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

71-74 °C (lit.)

SMILES string

[H]C(=O)c1ccc(OCc2ccccc2)cc1

InChI

1S/C14H12O2/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-10H,11H2

InChI key

ZVTWZSXLLMNMQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oriol Penon et al.
Journal of colloid and interface science, 462, 154-165 (2015-10-12)
The preparation of novel porphyrin derivatives and their immobilization onto iron oxide nanoparticles to build up suitable nanotools for potential use in photodynamic therapy (PDT) has been explored. To achieve this purpose, a zinc porphyrin derivative, ZnPR-COOH, has been synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service