Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900649

Sigma-Aldrich

D-Proline

Vetec, reagent grade, 98%

Synonym(s):

(R)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein/REAXYS Number:
80811
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

223 °C (dec.) (lit.)

SMILES string

OC(=O)[C@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m1/s1

InChI key

ONIBWKKTOPOVIA-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley C Campbell et al.
Archives of biochemistry and biophysics, 698, 108727-108727 (2020-12-18)
Proline utilization A (PutA) proteins are bifunctional proline catabolic enzymes that catalyze the 4-electron oxidation of l-proline to l-glutamate using spatially-separated proline dehydrogenase and l-glutamate-γ-semialdehyde dehydrogenase (GSALDH, a.k.a. ALDH4A1) active sites. The observation that l-proline inhibits both the GSALDH activity
Holly Newton et al.
Nature communications, 11(1), 4653-4653 (2020-09-18)
Cancer cells demand excess nutrients to support their proliferation, but how tumours exploit extracellular amino acids during systemic metabolic perturbations remain incompletely understood. Here, we use a Drosophila model of high-sugar diet (HSD)-enhanced tumourigenesis to uncover a systemic host-tumour metabolic
Yu-Sheng Shi et al.
Molecules (Basel, Switzerland), 20(6), 10793-10799 (2015-06-26)
A new diketopiperazine alkaloid named spirotryprostatin K (1), and five known alkaloids, spiro[5H,10H-dipyrrolo[1,2-a:1',2'-d]pyrazine-2(3H),2'-[2H]-indole]-3',5,10(1'H) trione (2), 6-methoxyspirotryprostatin B (3), pseurotin A (4), N-β-acetyltryptamine (5), and lumichrome (6) were isolated from the endophytic fungus Aspergillus fumigatus. The structure and the absolute configuration
Takahiro Takayama et al.
Analytica chimica acta, 898, 73-84 (2015-11-04)
Chiral metabolites are found in a wide variety of living organisms and some of them are understood to be physiologically active compounds and biomarkers. However, the overall analysis of chiral metabolomics is quite difficult due to the high number of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service