Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900643

Sigma-Aldrich

4-Nitroimidazole

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3O2
CAS Number:
Molecular Weight:
113.07
Beilstein/REAXYS Number:
2815
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

303 °C (dec.) (lit.)

SMILES string

[O-][N+](=O)c1c[nH]cn1

InChI

1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5)

InChI key

VYDWQPKRHOGLPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

392.0 °F - closed cup

flash_point_c

200 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taosheng Chen et al.
Assay and drug development technologies, 4(4), 387-396 (2006-09-02)
Receptor activator of nuclear factor-kappaB (NF-kappaB) (RANK) plays a key role in the differentiation, activation, and survival of osteoclasts. Upon activation of RANK with RANK ligand (RANKL), osteoclast precursor cells differentiate into tartrate-resistant acid phosphatase (TRAP)-positive, multinucleated osteoclasts. To identify
Thierry Juspin et al.
Molecules (Basel, Switzerland), 16(8), 6883-6993 (2011-08-16)
We report herein the synthesis of substituted 2-[4-(1,2-dimethyl-5-nitro-1H-imidazol-4-yl)phenyl]-1-arylethanols, ethyl 3-[4-(1,2-dimethyl-5-nitro-1H-imidazol-4-yl)-phenyl]-2-hydroxypropanoate and 2-[4-(1,2-dimethyl-5-nitro-1H-imidazol-4-yl)benzyl]-2-hydroxy-acenaphthylen-1(2H)-one from the reactions of 4-[4-(chloromethyl)phenyl]-1,2-dimethyl-5-nitro-1H-imidazole with various aromatic carbonyl and a-carbonyl ester derivatives using tetrakis(dimethylamino)ethylene (TDAE) methodology.
Soledad Fernández et al.
Bioorganic & medicinal chemistry, 20(13), 4040-4048 (2012-06-05)
An important issue in the development of metal-based radiopharmaceuticals is the selection of the labelling strategy in order to couple the metal to the pharmacophore without losing the biological activity. With the aim to evaluate the correlation between ligand denticity
Javier Giglio et al.
Bioorganic & medicinal chemistry letters, 21(1), 394-397 (2010-11-23)
With the aim to develop new potential (99m)Tc-radiopharmaceuticals for imaging hypoxia based on the formation of Tc-nitrido complexes, two novel dithiocarbamate containing metronidazole derivatives (L1 and L2) have been prepared and characterised. The synthesis of L1 and L2 was achieved
Soledad Fernández et al.
Nuclear medicine and biology, 40(2), 273-279 (2012-12-12)
Hypoxia imaging is an important field in radiopharmaceutical research since hypoxic cells are very resistant to radiation treatment and diffusional limitations restrict the efficacy of chemotherapy. Gallium-68 is a widely used radionuclide for positron emission tomography (PET) due to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service