Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900642

Sigma-Aldrich

2,6-Dihydroxybenzoic acid

Vetec, reagent grade, 98%

Synonym(s):

γ-Resorcylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein/REAXYS Number:
2209755
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

165 °C (dec.) (lit.)

SMILES string

OC(=O)c1c(O)cccc1O

InChI

1S/C7H6O4/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

AKEUNCKRJATALU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Shane Rountree et al.
Organic letters, 11(4), 871-874 (2009-01-28)
The facile preparation of a novel 8-membered polyhydroxylated salicylic acid lactone from 2,6-dihydroxybenzoic acid and sodium thio-D-glucose is described. The key step involved a sodium hydride promoted intramolecular lactonization in the presence of excess TMSCl, which led to isolation of
Derin Orhon et al.
Environmental technology, 33(13-15), 1517-1522 (2012-09-20)
The paper mainly focused on illustrating the merit of respirometric analysis in assessing the inhibitory/toxic impact of xenobiotics on substrate biodegradation. It also evaluated biodegradation characteristics of these chemicals at continuous exposure through acclimation ofthe microbial culture. The nature and
Tugce Katipoglu et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 45(14), 1885-1891 (2010-10-29)
This study evaluated the kinetics of 2,6-dihydroxybenozic acid and peptone biodegradation at low sludge age by acclimated culture under aerobic conditions. A laboratory-scale sequencing batch reactor was set and fed with peptone mixture. The system was operated at steady-state at
Masahiro Yoshida et al.
Journal of bacteriology, 189(5), 1573-1581 (2006-12-13)
We identified a gene cluster that is involved in the gamma-resorcylate (2,6-dihydroxybenzoate) catabolism of the aerobic bacterium Rhizobium sp. strain MTP-10005. The cluster consists of the graRDAFCBEK genes, and graA, graB, graC, and graD were heterologously expressed in Escherichia coli.
Toyokazu Yoshida et al.
Archives of microbiology, 181(6), 391-397 (2004-05-01)
A nonoxidative decarboxylase, 2,6-dihydroxybenzoate decarboxylase, was found in Agrobacterium tumefaciens IAM12048. The enzyme activity was induced specifically by 2,6-dihydroxybenzoate. The purified enzyme was a homotetramer of identical 38 kDa subunits. The purified decarboxylase catalyzed the nonoxidative decarboxylation of 2,6-dihydroxybenzoate and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service