Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900640

Sigma-Aldrich

1,2,4-Triazole

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3N3
CAS Number:
Molecular Weight:
69.07
Beilstein/REAXYS Number:
104767
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

260 °C (lit.)

mp

119-121 °C (lit.)

SMILES string

c1nc[nH]n1

InChI

1S/C2H3N3/c1-3-2-5-4-1/h1-2H,(H,3,4,5)

InChI key

NSPMIYGKQJPBQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

338.0 °F

flash_point_c

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Plech et al.
European journal of medicinal chemistry, 60, 128-134 (2013-01-05)
A series of novel 1,2,4-triazole-ciprofloxacin hybrids was designed, synthesised and evaluated in vitro against drug-susceptible and drug-resistant bacteria. A significant part of the compounds obtained showed antibacterial activity higher than the activity of ciprofloxacin, both towards Gram-positive and Gram-negative species.
Aline Moulin et al.
Amino acids, 44(2), 301-314 (2012-07-17)
Ghrelin is a 28-residue peptide acylated with an n-octanoyl group on the Ser 3 residue, predominantly produced by the stomach. Ghrelin displays strong growth hormone (GH) releasing activity, which is mediated by the activation of the so-called GH secretagogue receptor
Jolanta Flieger et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 909, 26-33 (2012-11-17)
This paper describes a novel reversed-phase high performance liquid chromatography (RP-HPLC) with photo-diode array detection (DAD) method for the determination of three new derivatives of 4-alkyl-5-(3-chlorophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione with different antiepileptic activity in the brains of mice treated with the doses of
Camila C Amorim et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(3), 183-190 (2013-01-30)
This study evaluated the adsorption capacity of ethylenthiourea (ETU) and 1H-1,2,4-triazole (1,2,4-T) for two commercial activated carbons: charcoal-powdered activated carbon (CPAC) and bovine bone-powdered activated carbon (BPAC). The tests were conducted at a bench scale, with ETU and 1,2,4-T diluted
Yaseen A Al-Soud et al.
Archiv der Pharmazie, 345(8), 610-621 (2012-04-26)
A series of disubstituted-1H-1,2,4-triazole derivatives was synthesized with the aim of developing new non-steroidal inhibitors of 17β-hydroxysteroid dehydrogenase type 2 (17βHSD2) - a novel and attractive target for the treatment of osteoporosis. 17βHSD2 catalyzes the oxidation of the highly active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service