Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900639

Sigma-Aldrich

Pyrazole

Vetec, reagent grade, 98%

Synonym(s):

1,2-Diazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein/REAXYS Number:
103775
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

bp

186-188 °C (lit.)

mp

67-70 °C (lit.)

SMILES string

c1cn[nH]c1

InChI

1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)

InChI key

WTKZEGDFNFYCGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1

target_organs

spleen,Thyroid

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javeed Ahmad Sheikh et al.
Inorganic chemistry, 52(17), 9717-9719 (2013-08-16)
A novel octadecanuclear copper pyrazolate-phosphonate nanocage with a bowl-shaped arrangement of the copper(II) centers in the asymmetric unit is reported. Characterization of intermediates in both solid and solution states aids to propose the mechanism of such a giant aggregation. Magnetic
Mohamed Jawed Ahsan et al.
Bioorganic & medicinal chemistry letters, 22(23), 7029-7035 (2012-10-27)
A series of 43, 3a,4-dihydro-3H-indeno[1,2-c]pyrazole-2-carboxamide/carbothioamide analogues (D01-D43) were analysed using Petra, Osiris, Molinspiration and ALOGPS (POMA) to identify pharmacophore, toxicity prediction, lipophilicity and bioactivity. All the compounds were evaluated for anti-HIV activity. 3-(4-Chlorophenyl)-N-(4-fluorophenyl)-6,7-dimethoxy-3a,4-dihydro-3H-indeno[1,2-c]pyrazole-2-carboxamide (D07) was found to be the most
Mingzhen Mao et al.
Bioorganic & medicinal chemistry letters, 23(1), 42-46 (2012-12-12)
In an attempt to search for potent insecticides targeting the ryanodine receptor (RyR), a series of novel N-pyridylpyrazolecarboxamides containing cyano substituent in the ortho-position were designed and synthesized. Their insecticidal activities of target compounds against oriental armyworm (Mythimna separata) and
James S Scott et al.
Journal of medicinal chemistry, 55(22), 10136-10147 (2012-10-24)
Inhibition of 11β-HSD1 is viewed as a potential target for the treatment of obesity and other elements of the metabolic syndrome. We report here the optimization of a carboxylic acid class of inhibitors from AZD4017 (1) to the development candidate
Xinfang Xu et al.
The Journal of organic chemistry, 78(4), 1583-1588 (2013-01-11)
A regiospecific synthesis of multifunctional pyrazoleshas been developed from a cascade process triggered by Rh(II)-catalyzed dinitrogen extrusion from enol diazoacetates with vinylogous nucleophilic addition followed by Lewis acid catalyzed cyclization and aromatization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service