Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900636

Sigma-Aldrich

Tetraphenylphosphonium bromide

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4P(Br)
CAS Number:
Molecular Weight:
419.29
Beilstein/REAXYS Number:
3922383
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

97%

mp

295-300 °C (lit.)

SMILES string

[Br-].c1ccc(cc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20P.BrH/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;1H/q+1;/p-1

InChI key

BRKFQVAOMSWFDU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhou et al.
Bioconjugate chemistry, 22(8), 1459-1472 (2011-06-24)
Alteration in mitochondrial transmembrane potential (ΔΨ(m)) is an important characteristic of cancer. The observation that the enhanced negative mitochondrial potential is prevalent in tumor cell phenotype provides a conceptual basis for development of mitochondrion-targeting therapeutic drugs and molecular imaging probes.
Denice C Bay et al.
Biochimica et biophysica acta, 1798(3), 526-535 (2009-12-29)
Escherichia coli multidrug resistance protein E (EmrE) is a four transmembrane alpha-helix protein, and a member of the small multidrug resistance protein family that confers resistance to a broad range of quaternary cation compounds (QCC) via proton motive force. The
Myocardial imaging for mitochondrial membrane potential.
H William Strauss et al.
JACC. Cardiovascular imaging, 5(3), 293-296 (2012-03-17)
M S El-Shahawi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(7), 757-757 (2011-07-13)
Fast and selective sorptions of Cr(VI) species from aqueous media onto tetraphenylphosphonium bromide (TPP(+) · Br(-)) physically immobilized polyurethane foams (PUFs) sorbent were achieved. Based on the Scatchard model of binding sites of the PUFs and Langmuir and Dubinin-Radushkevich (D-R)
Carlos M Palmeira et al.
Methods in molecular biology (Clifton, N.J.), 810, 89-101 (2011-11-08)
The study of the mitochondrial membrane potential (ΔΨ) is essential for an integrated appraisal of mitochondrial function, since it reflects differences in electrical potential and represents the main component of the proton electrochemical gradient accounting for more than 90% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service