Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900635

Sigma-Aldrich

2-Mercaptopyridine

Vetec, reagent grade, 98%

Synonym(s):

2-Pyridinethiol, 2-Pyridyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NS
CAS Number:
Molecular Weight:
111.16
Beilstein/REAXYS Number:
105787
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

127-130 °C (lit.)

storage temp.

2-8°C

SMILES string

Sc1ccccn1

InChI

1S/C5H5NS/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)

InChI key

WHMDPDGBKYUEMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gavin Dyson et al.
Dalton transactions (Cambridge, England : 2003), (31)(31), 6120-6126 (2010-05-08)
A new family of flexible scorpionate ligands based on 2-mercaptopyridine is reported. The tris- and bis-substituted ligands, K[HB(mp)(3)] (1) and Na[H(2)B(mp)(2)] (2) (mp = 2-mercaptopyridine) have been prepared and fully characterised. The structural characterisation of 1 reveals an unprecedented mu(3)-kappa(3)-SS'H-eta(1)eta(1)eta(2)-kappa(2)-S''C-eta(1)eta(1)-kappa(1)-S'-eta(1)
Sasa Koncarevic et al.
Free radical biology & medicine, 46(8), 1096-1108 (2009-05-15)
Terpyridineplatinum(II) complexes (TPCs) efficiently inhibit the proliferation of glioblastoma cells in vitro and have been tested successfully in a rodent glioblastoma model. Apart from intercalation with DNA, the major mechanism of action of TPCs is a very potent and specific
Emiliano Cortés et al.
Physical chemistry chemical physics : PCCP, 11(34), 7469-7475 (2009-08-20)
The engineering of cavity void metallic arrays allows to vary the plasmon-polariton mode energies from the near infrared to the ultraviolet through the tuning of the void height and diameter, and the selection of the appropriate material. Typically Au nanocavity
A M Attia et al.
Nucleosides & nucleotides, 17(8), 1355-1363 (1998-07-22)
The synthesis of new 4- and 5-substituted-3-cyanopyridine nucleosides has been performed by reacting the silylated pyridines and penta-omicron-acetyl-alpha -D-glycopyranose in dichloroethane in the presence of SnCl4. The free nucleosides were tested for their potential activity against HIV and different types
Jayaraman Narenkumar et al.
Chemosphere, 222, 611-618 (2019-02-08)
In this present study, the biocorrosion behaviour of Bacillus thuringiensis EN2 and B. oleronius EN9 on copper metal CW024A (Cu) in cooling water system (1% chloride) were evaluated using weight loss, electrochemical impedance spectroscopy (EIS) and surface analysis. In presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service