Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900631

Sigma-Aldrich

3′-Bromoacetophenone

Vetec, reagent grade, 98%

Synonym(s):

1-Acetyl-3-bromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCH3
CAS Number:
Molecular Weight:
199.04
Beilstein/REAXYS Number:
507599
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.576 (lit.)

mp

8-11 °C (lit.)

density

1.505 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccc(Br)c1

InChI

1S/C8H7BrO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,1H3

InChI key

JYAQYXOVOHJRCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service