Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900629

Sigma-Aldrich

4-Chlorophenylacetic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CO2H
CAS Number:
Molecular Weight:
170.59
Beilstein/REAXYS Number:
1072816
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

102-105 °C (lit.)

SMILES string

OC(=O)Cc1ccc(Cl)cc1

InChI

1S/C8H7ClO2/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

CDPKJZJVTHSESZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil Sidell et al.
Cancer letters, 251(2), 302-310 (2007-01-12)
Treatment of estrogen-sensitive breast cancer with selective estrogen selective modulators (SERMs) and, more recently, aromatase inhibitors has met with wide success. However, antagonism of estrogen receptor (ER) activity in breast carcinomas by SERMs such as tamoxifen has been associated with
S Sawatsri et al.
International journal of cancer, 93(5), 687-692 (2001-07-31)
The aromatic fatty acid phenylacetate (PA) and its analogs have come under intense investigation due to their ability to cause the growth arrest of a variety of neoplasia, including human breast cancer. We have determined that PA and its halide
N Sidell et al.
British journal of cancer, 89(2), 412-419 (2003-07-17)
We have investigated the effects of the low-toxic retinoid, all-trans retinoyl beta-glucuronide (RAG) alone and in combination with the phenylacetate (PA) derivative 4-chloro-phenylacetate (4-CPA) on the human neuroblastoma cell line, LA-N-5. In vitro studies demonstrated that RAG and 4-CPA treatments
A Markus et al.
Journal of bacteriology, 160(2), 618-621 (1984-11-01)
In cell-free extracts from Pseudomonas sp. strain CBS3 the conversion of 4-chlorophenylacetate to 3,4-dihydroxyphenylacetate was demonstrated. By Sephacryl S-200 chromatography two protein fractions, A and B, were obtained which both were essential for enzyme activity. Fe2+ and NADH were cofactors
A Markus et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 363(4), 431-437 (1982-04-01)
Pseudomonas spec. CBS 3 converts 4-chlorophenylacetic acid partly into 3-chloro-4-hydroxy-, 4-chloro-3-hydroxy-, and 4-chloro-2-hydroxyphenylacetic acid by the action of monooxygenases. However, these compounds are not intermediates in the degradation of 4-chlorophenylacetic acid. Pseudomonas spec. CBS 3 is not able to grow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service