Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900625

Sigma-Aldrich

γ-Cyclodextrin hydrate

Vetec, reagent grade

Synonym(s):

γ-Schardinger dextrin, Cyclooctaamylose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40 · xH2O
CAS Number:
Molecular Weight:
1297.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

reagent grade

product line

Vetec

mp

267 °C (dec.) (lit.)

SMILES string

O.OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]9[C@H](O)[C@@H](O)[C@H](O[C@@H]9CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C48H80O40.H2O/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58;/h9-72H,1-8H2;1H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-;/m1./s1

InChI key

SPKUKQHKKVTYOE-SMTXKKGASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Modifier used in experiments evaluating viability, integrity, and inflammation in genital epithelia upon exposure to pharmaceutical excipients and microbicides

Reactant used to investigate host-guest interactions between α−, β−and γ-cyclodextrins and vanadocene dichloride

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service