Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900623

Sigma-Aldrich

tert-Butyl acetoacetate

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein/REAXYS Number:
1680303
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

refractive index

n20/D 1.419 (lit.)

bp

71-72 °C/11 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)CC(=O)OC(C)(C)C

InChI

1S/C8H14O3/c1-6(9)5-7(10)11-8(2,3)4/h5H2,1-4H3

Inchi Key

JKUYRAMKJLMYLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

150.8 °F - closed cup

flash_point_c

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meghan C Drummond et al.
Nature communications, 6, 6873-6873 (2015-04-22)
The maintenance of sensory hair cell stereocilia is critical for lifelong hearing; however, mechanisms of structural homeostasis remain poorly understood. Conflicting models propose that stereocilia F-actin cores are either continually renewed every 24-48 h via a treadmill or are stable, exceptionally
Ying Wang et al.
Endocrinology, 155(5), 1970-1981 (2014-03-08)
FSH is an essential regulator of mammalian reproduction. Its synthesis by pituitary gonadotrope cells is regulated by multiple endocrine and paracrine factors, including TGFβ superfamily ligands, such as the activins and inhibins. Activins stimulate FSH synthesis via transcriptional regulation of
Hong Xie et al.
Oncotarget, 5(20), 10048-10057 (2014-10-11)
Telomerase activation through the induction of its catalytic component TERT is essential in carcinogenesis. The regulatory mechanism and clinical significance underlying cancer-specific TERT expression have been extensively investigated in various human malignancies, but little is known about these in Merkel
Jordan M Spatz et al.
The Journal of biological chemistry, 290(27), 16744-16758 (2015-05-09)
Although bone responds to its mechanical environment, the cellular and molecular mechanisms underlying the response of the skeleton to mechanical unloading are not completely understood. Osteocytes are the most abundant but least understood cells in bones and are thought to
Joseph G Crompton et al.
Cancer research, 75(2), 296-305 (2014-11-30)
Adoptive cell therapy (ACT) using autologous tumor-infiltrating lymphocytes (TIL) results in complete regression of advanced cancer in some patients, but the efficacy of this potentially curative therapy may be limited by poor persistence of TIL after adoptive transfer. Pharmacologic inhibition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service