Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900621

Sigma-Aldrich

4′-Ethoxyacetophenone

Vetec, reagent grade, 98%

Synonym(s):

1-(4-Ethoxyphenyl)ethan-1-one, 1-(4-Ethoxyphenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4COCH3
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

268-269 °C/758 mmHg (lit.)

mp

37-39 °C (lit.)

SMILES string

CCOc1ccc(cc1)C(C)=O

InChI

1S/C10H12O2/c1-3-12-10-6-4-9(5-7-10)8(2)11/h4-7H,3H2,1-2H3

InChI key

YJFNFQHMQJCPRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service