Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900616

Sigma-Aldrich

1,4-Dimethoxybenzene

Vetec, reagent grade, 98%

Synonym(s):

p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein/REAXYS Number:
774605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.8 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

Vetec

assay

98%

autoignition temp.

1463 °F

bp

213 °C (lit.)

mp

54-56 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC)cc1

InChI

1S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,1-2H3

InChI key

OHBQPCCCRFSCAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

257.0 °F - closed cup

flash_point_c

125 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Altmann et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 8, 114-116 (1985-01-01)
Butylated hydroxyanisole (BHA) is widely used as an antioxidant in foodstuffs, in materials which come into contact with food and also in cosmetic products. The safety of BHA was questioned, however, when it was reported that in a recent Japanese
M Masuda et al.
Bioscience, biotechnology, and biochemistry, 60(5), 806-810 (1996-05-01)
Bovine serum albumin (BSA) added to the reaction medium for the oxidative demethylation of 4-methoxyanisole and its "di-CD3" isotopomer ([d6]methoxyanisole), when catalyzed by liver microsomes from untreated rats, decreased the Km values and increased the V(max)/Km (= V/K) values. The
D W Kim et al.
Organic letters, 3(3), 445-447 (2001-06-29)
[figure: see text] Fused 1,4-dimethoxybenzenes could be oxidized to benzoquinones by either direct oxidation or demethylation-oxidation. The oxidative demethylation of 5,8-dimethoxy-2-methylquinoline using 1.1 equiv of NBS in aqueous THF and a catalytic amount of H2SO4 at 20 degrees C for
Tiantian Li et al.
The journal of physical chemistry. A, 115(19), 4988-4994 (2011-04-27)
The effect of substituents on the oxidation potential for the one-electron reaction of 1,4-dimethoxybenzene was understood with a theoretical calculation based on density functional theory (DFT) at the level of B3LYP/6-311+G(d). It is found that the oxidation potential for the
Stefan Dötterl et al.
Journal of chemical ecology, 31(12), 2993-2998 (2005-11-01)
Many bees are oligolectic and collect pollen for their larvae only from one particular plant family or genus. Here, we identified flower scent compounds of two Salix species important for the attraction of the oligolectic bee Andrena vaga, which collects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service