Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900615

Sigma-Aldrich

DL-Phenylalanine

Vetec, reagent grade, 98%

Synonym(s):

(±)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.19
Beilstein/REAXYS Number:
3198807
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

266-267 °C (dec.) (lit.)

SMILES string

NC(Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Søren Reitelseder et al.
Clinical nutrition (Edinburgh, Scotland), 39(12), 3652-3662 (2020-04-27)
Availability of dietary protein-derived amino acids (AA) is an important determinant for their utilization in metabolism and for protein synthesis. Intrinsic labeling of protein is the only method to directly trace availability and utilization. The purpose of the present study
Raja R Pillai et al.
The Journal of nutrition, 145(5), 954-959 (2015-03-13)
Lysine requirements of well-nourished children from developing regions have been found to be similar to those of children from developed regions (33.5 mg · kg⁻¹ · d⁻¹). However, intestinal parasites have been shown to increase lysine requirements in undernourished adults, and it is not
Sohei Nakayama et al.
Cancer research, 74(20), 5891-5902 (2014-08-29)
The discovery of somatic mutations in EGFR and development of EGFR tyrosine kinase inhibitors (TKI) have revolutionized treatment for lung cancer. However, resistance to TKIs emerges in almost all patients and currently no effective treatment is available. Here, we show
Katarzyna M Romek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8296-8301 (2015-06-25)
Tramadol, previously only known as a synthetic analgesic, has now been found in the bark and wood of roots of the African medicinal tree Nauclea latifolia. At present, no direct evidence is available as to the biosynthetic pathway of its
Rachel Schwartz-Narbonne et al.
Scientific reports, 5, 9791-9791 (2015-06-10)
Understanding woolly mammoth ecology is key to understanding Pleistocene community dynamics and evaluating the roles of human hunting and climate change in late Quaternary megafaunal extinctions. Previous isotopic studies of mammoths' diet and physiology have been hampered by the 'mammoth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service