Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900614

Sigma-Aldrich

4-Aminobenzoic acid

Vetec, reagent grade, 98%

Synonym(s):

PABA, Vitamin Bx, Vitamin H1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein/REAXYS Number:
471605
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

187-189 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

339.8 °F - closed cup

flash_point_c

171 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhou et al.
Water research, 47(1), 153-162 (2012-10-23)
Dissolved organic matter (DOM) is an important photosensitizer for the phototransformation of organic contaminants in sunlit natural waters. This article focuses on the photolysis kinetics and mechanism of sunscreen agent p-aminobenzoic acid (PABA) in the presence of four kinds of
Reza Forghani et al.
Radiology, 263(2), 451-460 (2012-03-23)
To evaluate myeloperoxidase (MPO) as a newer therapeutic target and bis-5-hydroxytryptamide-diethylenetriaminepentaacetate-gadolinium (Gd) (MPO-Gd) as an imaging biomarker for demyelinating diseases such as multiple sclerosis (MS) by using experimental autoimmune encephalomyelitis (EAE), a murine model of MS. Animal experiments were approved
L O Derewlany et al.
The Journal of pharmacology and experimental therapeutics, 269(2), 756-760 (1994-05-01)
The fetus is exposed to almost all of the substances found in the maternal circulation whether nutrients or foreign chemicals ("xenobiotics"). The main route of exposure is the placenta. The placenta is metabolically active toward xenobiotics and the nature of
Shu-Ting Chen et al.
Journal of the American Society for Mass Spectrometry, 23(8), 1408-1418 (2012-06-08)
A strategy based on negative ion electrospray ionization tandem mass spectrometry and closed-ring labeling with both 8-aminopyrene-1,3,6-trisulfonate (APTS) and p-aminobenzoic acid ethyl ester (ABEE) was developed for linkage and branch determination of high-mannose oligosaccharides. X-type cross-ring fragment ions obtained from
L O Derewlany et al.
The Journal of pharmacology and experimental therapeutics, 269(2), 761-765 (1994-05-01)
Studies in our laboratory have shown that the N-acetylation activity of the human term placenta is a predominantly attributable to the NAT1 form of arylamine N-acetyltransferase (NAT). To further assess the acetylation capacity of the placenta, the N-acetylation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service