Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900612

Sigma-Aldrich

2,6-Dimethoxybenzoic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein/REAXYS Number:
2212190
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

185-187 °C (lit.)

SMILES string

COc1cccc(OC)c1C(O)=O

InChI

1S/C9H10O4/c1-12-6-4-3-5-7(13-2)8(6)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

MBIZFBDREVRUHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Arrault et al.
Pest management science, 58(12), 1229-1235 (2002-12-13)
Nutrient-agar plates containing isoxaben (500 mg litre(-1)) were used to isolate isoxaben-metabolising bacteria from four European soils incubated with the herbicide under laboratory conditions. In flask experiments, inoculation of a basal salts medium containing nitrogen and [phenyl-U-14C]isoxaben with an isolate
Synthesis of aryl ketones by palladium(II)-catalyzed decarboxylative addition of benzoic acids to nitriles.
Jonas Lindh et al.
Angewandte Chemie (International ed. in English), 49(42), 7733-7737 (2010-09-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service