Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900605

Sigma-Aldrich

Diethyl adipate

Vetec, reagent grade, 98%

Synonym(s):

Hexanedioic acid 1,6-diethyl ester, NSC 19160, NSC 3363

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)4COOC2H5
CAS Number:
Molecular Weight:
202.25
Beilstein/REAXYS Number:
1780035
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.427 (lit.)

bp

251 °C (lit.)

mp

−20-−19 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCC(=O)OCC

InChI

1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3

InChI key

VIZORQUEIQEFRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronika Fischer et al.
Journal of colloid and interface science, 453, 186-193 (2015-05-20)
It was recently demonstrated that a nano-clustering was present in the monophasic "pre-Ouzo" region of ternary liquid mixtures without surfactants. The goal of this work is to check if this nano-clustering is also present in the surfactant-free and water-free "green"

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service