Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900604

Sigma-Aldrich

Ricinoleic acid

Vetec, reagent grade, 80%

Synonym(s):

(R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH2CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
298.46
Beilstein/REAXYS Number:
1727813
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

80%

density

0.940 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9-/t17-/m1/s1

InChI key

WBHHMMIMDMUBKC-QJWNTBNXSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

435.2 °F - closed cup

flash_point_c

224 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan L Robertson et al.
ACS applied materials & interfaces, 3(9), 3402-3410 (2011-08-10)
Poly(l-lactide) (PLLA) is a renewable resource polymer derived from plant sugars with several commercial applications. Broader implementation of the material is limited due to its inherent brittleness. We show that the addition of 5 wt % castor oil to PLLA
Petra Kocbek et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 50(1), 130-138 (2013-04-23)
Superparamagnetic iron oxide nanoparticles (SPIONs) show a great promise for a wide specter of bioapplications, due to their characteristic magnetic properties exhibited only in the presence of magnetic field. Their advantages in the fields of magnetic drug targeting and imaging
Ming Li Wang et al.
Journal of agricultural and food chemistry, 59(17), 9250-9256 (2011-08-16)
Castor has tremendous potential as a feedstock for biodiesel production. The oil content and fatty acid composition in castor seed are important factors determining the price for production and affecting the key fuel properties of biodiesel. There are 1033 available
Bruno Rodrigues Sampieri et al.
Microscopy research and technique, 75(5), 683-690 (2011-12-02)
Rhipicephalus sanguineus is a widely distributed tick species that has adapted to the urban environment, and the dog is its main host. This species is also known as a vector and reservoir of diseases caused by bacteria, protozoa, and viruses.
Adrian P Brown et al.
PloS one, 7(2), e30100-e30100 (2012-02-10)
Storage triacylglycerols in castor bean seeds are enriched in the hydroxylated fatty acid ricinoleate. Extensive tissue-specific RNA-Seq transcriptome and lipid analysis will help identify components important for its biosynthesis. Storage triacylglycerols (TAGs) in the endosperm of developing castor (Ricinus communis)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service