Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900601

Sigma-Aldrich

Methyl 2-aminobenzoate

Vetec, reagent grade, 98%

Synonym(s):

Methyl anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(H2N)C6H4CO2CH3
CAS Number:
Molecular Weight:
151.16
Beilstein/REAXYS Number:
606965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

1 mmHg ( 20 °C)

product line

Vetec

assay

98%

autoignition temp.

986 °F

refractive index

n20/D 1.582 (lit.)

bp

256 °C (lit.)

mp

24 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1N

InChI

1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3

InChI key

VAMXMNNIEUEQDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris E Houlden et al.
Angewandte Chemie (International ed. in English), 48(10), 1830-1833 (2009-01-27)
Pd and CO--ureally got me! The title reaction proceeds efficiently at 18 degrees C under CO (1 atm) with 5 % [Pd(OTs)(2)(MeCN)(2)] as precatalyst. Depending on the solvents used, either anthranilates or cyclic imides can be obtained in high yields
N R Lien et al.
Carbohydrate research, 344(18), 2606-2608 (2009-11-03)
A 2:1 complex between cyclomaltoheptaose (beta-cyclodextrin) and N-methylanthranilic acid has been studied in the solid state. The inclusion complex belongs to the triclinic system (space group P1) with unit cell dimensions a=15.2773(15)A, b=15.4710(15)A, c=17.9627(18)A, alpha=99.632(5) degrees , beta=113.416(5) degrees
Marcus Taupp et al.
Journal of agricultural and food chemistry, 53(24), 9586-9589 (2005-11-24)
Bacillus megaterium, isolated in a screening process from topsoil, was used for N-demethylation of natural N-methyl methyl anthranilate to produce natural methyl anthranilate. Maximal productivity of 70 mg/L/day was achieved under laboratory-scale conditions without further optimization. No byproducts were observed.
Yuriko Ito et al.
Journal of agricultural and food chemistry, 50(17), 4878-4884 (2002-08-09)
The odorants in Chinese jasmine green tea scented with jasmine flowers (Jasminum sambac) were separated from the infusion by adsorption to Porapak Q resin. Among the 66 compounds identified by GC and GC/MS, linalool (floral), methyl anthranilate (grape-like), 4-hexanolide (sweet)
S Moran
Pest management science, 57(3), 248-252 (2001-07-18)
Two methyl anthranilate formulations, ReJex-iT TP-40 and AG-36, were tested as bird-repelling agents for animal feed and vegetable sprouts, respectively. Feral pigeons (Columbia livia) and house sparrows (Passer domesticus) are important pests of animal husbandry and field crops in Israel.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service