Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900597

Sigma-Aldrich

Diphenyl sulfone

Vetec, reagent grade

Synonym(s):

Phenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2SO2
CAS Number:
Molecular Weight:
218.27
Beilstein/REAXYS Number:
1910573
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

bp

379 °C (lit.)

mp

123-129 °C (lit.)

SMILES string

O=S(=O)(c1ccccc1)c2ccccc2

InChI

1S/C12H10O2S/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

KZTYYGOKRVBIMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

363.2 °F

flash_point_c

184 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service