Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900595

Sigma-Aldrich

Azelaic acid

Vetec, reagent grade

Synonym(s):

Nonanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)7CO2H
CAS Number:
Molecular Weight:
188.22
Beilstein/REAXYS Number:
1101094
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

6.5 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

Vetec

bp

286 °C/100 mmHg (lit.)

mp

109-111 °C (lit.)

SMILES string

OC(=O)CCCCCCCC(O)=O

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)

InChI key

BDJRBEYXGGNYIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

410.0 °F - closed cup

flash_point_c

210 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alvin B Coda et al.
Journal of the American Academy of Dermatology, 69(4), 570-577 (2013-07-23)
Excess cathelicidin and kallikrein 5 (KLK5) have been hypothesized to play a role in the pathophysiology of rosacea. We sought to evaluate the effects of azelaic acid (AzA) on these elements of the innate immune system. Gene expression and protease
Esther J van Zuuren et al.
The Cochrane database of systematic reviews, (3)(3), CD003262-CD003262 (2011-03-18)
Rosacea is a common chronic skin condition affecting the face, characterised by flushing, redness, pimples, pustules, and dilated blood vessels. The eyes are often involved and thickening of the skin with enlargement (phymas), especially of the nose, can occur in
Maria Vinaixa et al.
PloS one, 6(12), e29052-e29052 (2011-12-24)
Polycystic ovary syndrome (PCOS) is a variable disorder characterized by a broad spectrum of anomalies, including hyperandrogenemia, insulin resistance, dyslipidemia, body adiposity, low-grade inflammation and increased cardiovascular disease risks. Recently, a new polytherapy consisting of low-dose flutamide, metformin and pioglitazone
Esther J van Zuuren et al.
Journal of the American Academy of Dermatology, 56(1), 107-115 (2006-12-28)
Rosacea is a common chronic skin and ocular condition. It is unclear which treatments are most effective. We have conducted a Cochrane review of rosacea therapies. This article is a distillation of that work. We sought to assess the evidence
J F Hermanns et al.
Dermatology (Basel, Switzerland), 201(2), 118-122 (2000-10-29)
During photoaging, the density of melanin chromatophores is heterogeneous in the epidermis. To define the patterns of pheomelanin-enriched melanotic hypermelanosis of the face in phototype II subjects and to assess the effect of depigmenting agents. Azelaic acid and glycolic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service