Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900593

Sigma-Aldrich

Diethyl succinate

Vetec, reagent grade, 98%

Synonym(s):

1,4-Diethyl butanedioate, Diethyl butanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
Beilstein/REAXYS Number:
907645
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

6 (vs air)

product line

Vetec

assay

98%

refractive index

n20/D 1.42 (lit.)

bp

218 °C (lit.)

mp

−20 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCC(=O)OCC

InChI

1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3

Inchi Key

DKMROQRQHGEIOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhu Xu et al.
Journal of texture studies, 52(1), 124-136 (2020-11-14)
Drinking tastes and lubrication properties of Chinese rice wine (CRW) under different heating temperatures were studied by tribology tests, gas chromatography-mass spectrometry (GC-MS) and sensory evaluations. CRW's drinking tastes were evaluated by taste panelists. Flavor compounds were detected by GC-MS.
Florian Diot-Néant et al.
ChemSusChem, 13(10), 2613-2620 (2020-04-03)
The already-reported, low-yielding, and non-sustainable Et3 N-mediated homocoupling of levoglucosenone (LGO) into the corresponding LGO-CyreneTM diketone has been revisited and greened-up. The use of methanol as both a renewable solvent and catalyst and K2 CO3 as a safe inorganic base
Niki M Zacharias et al.
Journal of the American Chemical Society, 134(2), 934-943 (2011-12-08)
The Krebs tricarboxylic acid cycle (TCA) is central to metabolic energy production and is known to be altered in many disease states. Real-time molecular imaging of the TCA cycle in vivo will be important in understanding the metabolic basis of
M B Ashour et al.
Toxicology and applied pharmacology, 89(3), 361-369 (1987-07-01)
Treatment with 0.5% (w/w) dietary clofibrate, a peroxisome proliferator, for 14 days induced microsomal carboxylesterase activities for five substrates including malathion, clofibrate, diethylsuccinate, diethylphthalate, and p-nitrophenylacetate in liver and kidney of male Swiss-Webster mice and Sprague-Dawley rats. The induction was
M B Ashour et al.
Biochemical pharmacology, 36(12), 1869-1879 (1987-06-15)
A series of substituted trifluoroketones were tested as inhibitors of mammalian liver microsomal carboxylesterase(s) hydrolyzing a variety of substrates including malathion, diethylsuccinate (DES) and p-nitrophenyl acetate (p-NpAc). The trifluoroketones used were very potent "transition state" inhibitors of crude mouse and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service