Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900592

Sigma-Aldrich

4-Hydroxybenzaldehyde

Vetec, reagent grade, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
471352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

95%

mp

112-116 °C (lit.)

SMILES string

[H]C(=O)c1ccc(O)cc1

InChI

1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H

Inchi Key

RGHHSNMVTDWUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

345.2 °F

flash_point_c

174 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhentian Lei et al.
Journal of proteome research, 9(12), 6220-6231 (2010-09-22)
The temporal proteome response of Medicago truncatula suspension cell cultures to yeast elicitation (which mimics fungal infection) was investigated using two-dimensional polyacrylamide gel electrophoresis (2-DE) and nanoliquid chromatography coupled to tandem mass spectrometry (nano LC-MS/MS). Reproducibility of 2-DE was assessed
Xinyi Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(31), 3297-3302 (2010-11-09)
A sensitive and reproducible HPLC method for quantitative determination of puerarin (PUE) in rat plasma was developed and validated using 4-hydroxybenzaldehyde as an internal standard. The separation of PUE was performed on a CAPCELL PAK C18 column by gradient elution
Nadja Schultz-Jensen et al.
Applied biochemistry and biotechnology, 165(3-4), 1010-1023 (2011-07-06)
The potential of wheat straw for ethanol production after pretreatment with O(3) generated in a plasma at atmospheric pressure and room temperature followed by fermentation was investigated. We found that cellulose and hemicellulose remained unaltered after ozonisation and a subsequent
Naoki Okamoto et al.
Journal of biochemistry, 150(5), 563-568 (2011-08-16)
Old yellow enzyme (OYE) is an NADPH oxidoreductase capable of reducing a variety of compounds. It contains flavin mononucleotide (FMN) as a prosthetic group. A ternary complex structure of OYE from Trypanosoma cruzi (TcOYE) with FMN and one of the
Ben M Goff et al.
Journal of the science of food and agriculture, 91(8), 1523-1526 (2011-05-05)
Sorghum [Sorghum bicolor (L.) Moench] has been shown to contain the cyanogenic glycoside dhurrin, which is responsible for the disorder known as prussic acid poisoning in livestock. The current standard method for estimating hydrogen cyanide (HCN) uses spectrophotometry to measure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service