V900590
3-Nitrobenzoic acid
Vetec™, reagent grade, 98%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
O2NC6H4CO2H
CAS Number:
Molecular Weight:
167.12
Beilstein/REAXYS Number:
908644
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
reagent grade
product line
Vetec™
assay
98%
mp
139-141 °C (lit.)
SMILES string
OC(=O)c1cccc(c1)[N+]([O-])=O
InChI
1S/C7H5NO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)
InChI key
AFPHTEQTJZKQAQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
374.0 °F - closed cup
flash_point_c
190 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhiqiang Hu et al.
Water research, 39(15), 3501-3510 (2005-07-30)
Biodegradation rates of benzoate and related aromatic compounds, 3-nitrobenzoate, 4-chlorobenzoate, 4-chlorophenol, and 2,4-dichlorophenol by unexposed (unacclimated) and long-term exposed (acclimated) biomass were quantified using a modified fed-batch technique. The acclimated biomass was taken after approximately 1-year of operation from three
L J Nadeau et al.
Applied and environmental microbiology, 61(2), 840-843 (1995-02-01)
Pseudomonas sp. strain JS51 grows on m-nitrobenzoate (m-NBA) with stoichiometric release of nitrite. m-NBA-grown cells oxidized m-NBA and protocatechuate but not 3-hydroxybenzoate, 4-hydroxy-3-nitrobenzoate, 4-nitrocatechol, and 1,2,4-benzenetriol. Protocatechuate accumulated transiently when succinate-grown cells were transferred to media containing m-NBA. Respirometric experiments
C R Bloom et al.
Biochemistry, 37(31), 10937-10944 (1998-08-07)
The positive and negative cooperativity and apparent half-site reactivity of the Co(II)-substituted insulin hexamer are well-described by a three-state allosteric model involving ligand-mediated interconversions between the three states: T3T3' right harpoon over left harpoon T3o R3o right harpoon over left
M Samsonowicz et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 82(1), 235-246 (2011-08-25)
The influence of beryllium, magnesium, calcium, strontium and barium cations on the electronic system of 3-nitrobenzoic acid was studied in comparison with studied earlier alkali metal ions. The vibrational FT-IR (in KBr and ATR techniques) and (1)H and (13)C NMR
Reproductive toxicology. m-Nitrobenzoic acid.
Environmental health perspectives, 105 Suppl 1, 325-326 (1997-02-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service