Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900588

Sigma-Aldrich

Vanillic acid

Vetec, reagent grade, 97%

Synonym(s):

4-Hydroxy-3-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein/REAXYS Number:
2208364
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

208-210 °C (lit.)

SMILES string

COc1cc(ccc1O)C(O)=O

InChI

1S/C8H8O4/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,9H,1H3,(H,10,11)

InChI key

WKOLLVMJNQIZCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael Llorach et al.
Journal of proteome research, 8(11), 5060-5068 (2009-09-17)
Cocoa-phytochemicals have been related to the health-benefits of cocoa consumption. Metabolomics has been proposed as a powerful tool to characterize both the intake and the effects on the metabolism of dietary components. Human urine metabolome modifications after single cocoa intake
B L Lee et al.
Clinical chemistry, 39(9), 1788-1792 (1993-09-01)
We describe a sensitive and specific high-performance liquid-chromatographic method for determining the benzene metabolite, trans,trans-muconic acid (ttMA) in urine by measuring ultraviolet absorbance at 265 nm. We mix 1 mL of urine sample with 2 mL of Tris buffer containing
F A Muskiet et al.
Clinical chemistry, 25(7), 1281-1284 (1979-07-01)
We report quantitative data on beta-glucuronidase- and sulfatase-hydrolyzable conjugates of homovanillic acid, 3,4-dihydroxyphenylacetic acid, p-hydroxyphenylacetic acid, and vanillic acid in the urine of 20 apparently normal and healthy control persons and of three patients with neuroblastoma. We used organic solvent
Changgeng Liu et al.
Environmental science & technology, 46(24), 13262-13269 (2012-11-23)
Methoxyphenols, tracers for wood smoke, are emitted into the atmosphere in large quantities, but their chemical degradation in the atmosphere has not been well characterized. In this study, heterogeneous kinetics of particulate syringaldehyde (SA), vanillic acid (VA), and coniferyl aldehyde
F Mashige et al.
Journal of chromatography. B, Biomedical applications, 658(1), 63-68 (1994-08-05)
An HPLC system for the simultaneous determination of acidic catecholamine metabolites, related compounds and 5-hydroxyindoleacetic acid (5-HIAA) in human urine was developed. A mixed-mode (C18/anion-exchange) column with isocratic elution using citrate buffer and an eight-channel electrochemical detector were used. Vanilmandelic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service