Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900587

Sigma-Aldrich

3-Ethoxy-4-hydroxybenzaldehyde

Vetec, reagent grade, 99%

Synonym(s):

Ethylvanillin, NSC 1803, NSC 67240

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(OH)CHO
CAS Number:
Molecular Weight:
166.17
Beilstein/REAXYS Number:
1073761
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

<0.01 mmHg ( 25 °C)

product line

Vetec

assay

99%

mp

74-77 °C (lit.)

SMILES string

[H]C(=O)c1ccc(O)c(OCC)c1

InChI

1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3

InChI key

CBOQJANXLMLOSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

293.0 °F - closed cup

flash_point_c

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Joo Jung et al.
Archives of pharmacal research, 33(2), 309-316 (2010-03-03)
The present work aimed to assess novel pharmacological properties of ethyl vanillin (EVA) which is used as a flavoring agent for cakes, dessert, confectionary, etc. EVA exhibited an inhibitory activity in the chorioallantoic membrane angiogenesis. Anti-inflammatory activity of EVA was
P Duchamp-Viret et al.
Science (New York, N.Y.), 284(5423), 2171-2174 (1999-06-26)
Molecular biology studies of olfaction have identified a multigene family of molecular receptors that are likely to be involved in odor transduction mechanisms. However, because previous functional data on peripheral coding were mainly collected from inferior vertebrates, it has been
E Jagerdeo et al.
Journal of AOAC International, 83(1), 237-240 (2000-02-29)
This paper describes a reversed-phase liquid chromatographic method for the determination of vanillin, associated natural aromatic compounds and/or synthetic precursors, ethyl vanillin, and coumarin, a commonly encountered adulterant in nonbeverage and beverage alcohol products using a ternary gradient mobile phase.
Mónica Avila et al.
Electrophoresis, 30(19), 3413-3418 (2009-09-10)
A new strategy based on the fast separation of the fingerprint markers of Vanilla planifolia extracts and vanilla-related samples on microfluidic-electrochemistry chip is proposed. This methodology allowed the detection of all required markers for confirmation of common frauds in this
Gökçe Polat Yemiş et al.
Journal of food protection, 74(12), 2062-2069 (2011-12-22)
Preservatives could be part of an effective intervention strategy for the control of Cronobacter species in foods, but few compounds with the desired antimicrobial properties have been identified to date. We examined the antibacterial activity of vanillin, ethyl vanillin, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service