Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900586

Sigma-Aldrich

Dimethyl terephthalate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein/REAXYS Number:
1107185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

1.04 (vs air)

vapor pressure

1.15 mmHg ( 93 °C)

product line

Vetec

assay

98%

autoignition temp.

1058 °F

mp

139-141 °C (lit.)

SMILES string

COC(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3

InChI key

WOZVHXUHUFLZGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

285.8 °F - closed cup

flash_point_c

141 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiqiang Guo et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(2), 337-340 (2007-02-28)
Two 10-hydroxybenzo[h]quinoline metal complexes, bis(10-hydroxybenzo[h]quinolinato) beryllium (Bebq2) and bis(10-hydroxybenzo[h]quinolinato)zincum (Znbq2), have been synthesized. The structure are characterized by 1HNMR, IR and so on. The photophysical processes of Bebq2 and Znbq2 have been carefully investigated by fluorescence spectra. The results show
Minoru Genta et al.
Waste management (New York, N.Y.), 27(9), 1167-1177 (2006-08-18)
To apply PET depolymerization in supercritical methanol to commercial recycling, the benefits of supercritical methanol usage in PET depolymerization was investigated from the viewpoint of the reaction rate and energy demands. PET was depolymerized in a batch reactor at 573
S Monarca et al.
Mutation research, 262(2), 85-92 (1991-02-01)
Dimethyl terephthalate (DMTP), the para configuration of dimethyl phthalate, is one of the basic monomers used in the synthesis of polyethylene terephthalate (PET) plastics. Human exposure to DMTP may primarily occur during the manufacture of PET fibers and films. The
R I Goncharova et al.
Genetika, 24(7), 1226-1233 (1988-07-01)
Mutagenic activity of dimethyl terephthalate (DMtP) was evaluated using the bone marrow micronucleus test in mice. Clear clastogenic effect with the highest response in 24 h after a single i.p. injection was obtained at all concentrations used (0.2-1.0 mM/kg). The
R I Goncharova et al.
TSitologiia i genetika, 36(1), 14-25 (2002-05-16)
In the mouse transplacental test, EMS induced micronuclei and disturbed haemopoiesis in female bone marrow and foetal liver. Dimethyl terephthalate at the tested dose was ineffective in pregnant females increasing however the level of these events in foetuses. Hence, both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service