Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900585

Sigma-Aldrich

4-Aminophenylacetic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein/REAXYS Number:
2208094
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to yellow

mp

201 °C (dec.) (lit.)

SMILES string

Nc1ccc(CC(O)=O)cc1

InChI

1S/C8H9NO2/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4H,5,9H2,(H,10,11)

InChI key

CSEWAUGPAQPMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed H Bedair et al.
Acta pharmaceutica (Zagreb, Croatia), 56(3), 273-284 (2006-09-01)
Condensation of 4-aminophenylacetic acid with phthalic anhydride gave (dioxoisoindolin-2-yl)phenylacetic acid (1), which was employed as the key intermediate in the synthesis of title compounds 2-8. The products were characterized by analytical and spectral data (IR, 1H NMR, 13C NMR and
C S Temple et al.
The Journal of biological chemistry, 273(1), 20-22 (1998-02-07)
4-Aminophenylacetic acid (4-APAA), a peptide mimic lacking a peptide bond, has been shown to interact with a proton-coupled oligopeptide transporter using a number of different experimental approaches. In addition to inhibiting transport of labeled peptides, these studies show that 4-APAA
D Meredith et al.
The Journal of physiology, 512 ( Pt 3), 629-634 (1999-01-09)
1. 4-Aminomethylbenzoic acid, a molecule which mimics the special configuration of a dipeptide, competitively inhibits peptide influx in both Xenopus Laevis oocytes expressing rabbit PepT1 and through PepT1 in rat renal brush border membrane vesicles. 2. This molecule is not
Douglas C McVey et al.
Digestive diseases and sciences, 50(3), 565-573 (2005-04-07)
A new compound, APAZA, consisting of a molecule of 5-aminosalicylic acid linked to one molecule of 4-aminophenylacetic acid by an azo bond, was testedfor its ability to inhibit acute colitis in rats caused by Clostridium difficile toxin A. When administered
Hongye Zhang et al.
Journal of colloid and interface science, 338(2), 468-473 (2009-07-21)
Monodispersed TiO2 hybrid microspheres were prepared via the hydrolysis of titanium isopropoxide (TTIP) in ethanol solution containing p-aminophenylacetic acid (APA). The effects of the APA:TTIP molar ratio, water content, reaction time and reaction temperature on the morphology of the resultant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service