Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900582

Sigma-Aldrich

2′-Hydroxyacetophenone

Vetec, reagent grade, 98%

Synonym(s):

2-Acetylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Beilstein/REAXYS Number:
386123
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.7 (vs air)

vapor pressure

~0.2 mmHg ( 20 °C)

product line

Vetec

assay

98%

refractive index

n20/D 1.558 (lit.)

bp

213 °C/717 mmHg (lit.)

mp

3-6 °C (lit.)

density

1.133 g/mL at 20 °C
1.131 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccccc1O

InChI

1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3

InChI key

JECYUBVRTQDVAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniele Gabriel et al.
Journal of cardiovascular pharmacology, 57(1), 20-27 (2011-01-15)
Isatin (1H-indole-2,3 dione) is an endogenous compound with biological activities. Many of its derivatives have pharmacological effects, including inhibition of cyclic guanosine monophosphate levels in cardiac tissue; sedative-hypnotic profiles; anticonvulsant, analgesic, antithermic, and anti-inflammatory activities; and anxiolytic, antimicrobial, and proapoptotic
Nora H Al-Shaalan
Molecules (Basel, Switzerland), 16(10), 8629-8645 (2011-10-15)
The Schiff base hydrazone ligand HL was prepared by the condensation reaction of 7-chloro-4-quinoline with o-hydroxyacetophenone. The ligand behaves either as monobasic bidentate or dibasic tridentate and contain ONN coordination sites. This was accounted for be the presence in the
Mohammed Y Merza et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(7), 1677-1683 (2004-05-19)
Ultra violet absorption spectra of o-methylacetophenone, o-fluoroacetophenone and o-hydroxyacetophenone solutions in different solvents are recorded in the region 200-350 nm at room temperature. Excited state dipole moments for three (pi* <-- pi) transitions of the benzene ring for solutions in
Xiao-Feng Wu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(40), 12595-12598 (2012-08-29)
Flavone of the month: a general and efficient method for the palladium-catalyzed carbonylative synthesis of flavones has been developed. Starting from aryl bromides and 2-hydroxyacetophenones, the corresponding flavones have been isolated in good yields.
R R Zaky et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 683-694 (2012-08-14)
The o-Hydroxy acetophenone [N-(3-hydroxy-2-naphthoyl)] hydrazone (H(2)o-HAHNH) has been prepared and its structure is confirmed by elemental analysis, IR, (1)H NMR and (13)C NMR spectroscopy. It has been used to produce diverse complexes with Co(II), Cd(II), Hg(II) and U(VI)O(2) ions. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service