Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900578

Sigma-Aldrich

Phenyl salicylate

Vetec, reagent grade, 98%

Synonym(s):

Salol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
Beilstein/REAXYS Number:
393969
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

liquid crystal

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Baran et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 81(3 Pt 1), 031503-031503 (2010-04-07)
We report the investigation of glass-forming salol upon different courses of the temperature changes from liquid to glass state and back using FT-IR spectroscopy measurements in the wide spectral and temperature ranges. The formation of the ordered clusters in supercooled
Cross-sensitivity between resorcinol, resorcinol monobenzoate and phenyl salicylate.
M Nakagawa et al.
Contact dermatitis, 27(3), 199-200 (1992-09-01)
P T Deota et al.
Natural product research, 17(1), 21-26 (2003-04-04)
The effect of photostabilization of azadirachtin-A (Aza-A) was examined in solutions when exposed to UV radiation, in the presence of four structurally different UV absorbers namely, p-aminobenzoic acid, 2,4-dihydroxybenzophenone, 4,4'-dihydroxybenzophenone and phenyl salicylate. The percentages of Aza-A recovered from the
C D Calnan et al.
Contact dermatitis, 7(4), 208-211 (1981-07-01)
Six cases of contact dermatitis from a lip salve are described. Five were allergic to phenyl salicylate and one to geraniol in the fragrance. The dermatitis spread in a ring around the mouth. Phenyl salicylate has been removed from the
Allergic contact dermatitis to various salols (phenyl salicylates). A structure-activity relationship study in man and in animal (guinea pig).
B Marchand et al.
Archives of dermatological research, 272(1-2), 61-66 (1982-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service