Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900575

Sigma-Aldrich

2,2,2-Trichloroethanol

Vetec, reagent grade, 98%

Synonym(s):

Trichloroethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH2OH
CAS Number:
Molecular Weight:
149.40
Beilstein/REAXYS Number:
1697495
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.16 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

Vetec

assay

98%

refractive index

n20/D 1.49 (lit.)

bp

152-154 °C (lit.)

mp

17.8 °C (lit.)

density

1.56 g/mL at 20 °C (lit.)

SMILES string

OCC(Cl)(Cl)Cl

InChI

1S/C2H3Cl3O/c3-2(4,5)1-6/h6H,1H2

InChI key

KPWDGTGXUYRARH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol L Ladner et al.
Analytical chemistry, 78(7), 2388-2396 (2006-04-04)
Proteins visualized by 2,2,2-trichloroethanol (TCE) on two-dimensional electrophoresis gels are efficiently identified by matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry (MS) and MS/MS. In a previous study, a method was developed that placed TCE in the polyacrylamide gel so that
Gavin Harrold et al.
Journal of contaminant hydrology, 80(1-2), 1-17 (2005-08-16)
The wettability of aquifer rocks is a key physical parameter which exerts an important control on the transport, residual trapping, distribution and eventual fate of chlorinated hydrocarbon solvents (CHSs) released into the subsurface. Typically chlorinated solvents are assumed to be
Xiang-Qun Hu et al.
British journal of pharmacology, 148(1), 88-101 (2006-03-08)
1 The goal of this study was to determine whether mutations of L293 at the 15' position of TM2 in the 5-HT(3A) receptor alter macroscopic current kinetics, and if these changes could account for alterations in alcohol modulation. Receptor function
Mariella Carrieri et al.
Archives of toxicology, 81(7), 529-532 (2007-02-08)
Nonfatal acute inhalation of trichloroethylene (TRI) at work was described. The subject, male, 54 years old, was drawn unconscious by a metal-degreasing machine and immediately sheltered in intensive care unit. Other than basic life support and common laboratory indices, blood
Apryl Bronley-DeLancey et al.
Environmental health perspectives, 114(8), 1237-1242 (2006-08-03)
Trichloroethylene (TCE) is a suspected human carcinogen and a common groundwater contaminant. Chloral hydrate (CH) is the major metabolite of TCE formed in the liver by cytochrome P450 2E1. CH is metabolized to the hepatocarcinogen trichloroacetate (TCA) by aldehyde dehydrogenase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service