Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900574

Sigma-Aldrich

1-Octadecanol

Vetec, reagent grade, 94%

Synonym(s):

Octadecyl alcohol, Stearyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OH
CAS Number:
Molecular Weight:
270.49
Beilstein/REAXYS Number:
1362907
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

9.3 (vs air)

vapor pressure

<0.01 mmHg ( 38 °C)

product line

Vetec

assay

94%

autoignition temp.

842 °F

expl. lim.

~8 %

mp

56-59 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCO

InChI

1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

GLDOVTGHNKAZLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

383.0 °F - closed cup

flash_point_c

195 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvie Roke et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(36), 13310-13314 (2006-08-30)
We investigate the phase behavior of surface-functionalized silica colloids at both the molecular and macroscopic levels. This investigation allows us to relate collective properties such as aggregation, gelation, and aging directly to molecular interfacial behavior. By using surface-specific vibrational spectroscopy
A Antonelli et al.
Nanotechnology, 21(42), 425101-425101 (2010-09-23)
Single-walled carbon nanotubes (SWNTs) due to their unique structural and physicochemical properties, have been proposed as delivery systems for a variety of diagnostic and therapeutic agents. However, SWNTs have proven difficult to solubilize in aqueous solution, limiting their use in
Laura Carreto et al.
Biophysical journal, 82(1 Pt 1), 530-540 (2001-12-26)
As a model for the epicuticular wax layer of plant cuticular membranes, we have studied the phase behavior of 1-tetradecanol and 1-octadecanol and their binary mixtures between 5 and 70 degrees C, using differential scanning calorimetry and Fourier transform infrared
M D Koone et al.
Archives of dermatology, 126(11), 1485-1490 (1990-11-01)
A number of inherited disorders of cornification have been related to abnormal lipid metabolism. In the recessively inherited Sjögren-Larsson syndrome, defined by the triad of ichthyosis, mental retardation, and spasticity, fatty alcohol:NAD+ oxidoreductase deficiency has recently been reported. These patients
Jörg Bauer et al.
Journal of the American Chemical Society, 127(20), 7296-7297 (2005-05-19)
Hydrophobically assisted switching phase (HASP) synthesis is a concept that allows the choice between the advantages of solid-supported chemistry and those of solution-phase synthesis. Starting from the examination of adsorption and desorption properties of hydrophobic molecules to and from reversed-phase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service