Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900573

Sigma-Aldrich

1-Bromooctadecane

Vetec, reagent grade, 97%

Synonym(s):

Octadecyl bromide, Stearyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17Br
CAS Number:
Molecular Weight:
333.39
Beilstein/REAXYS Number:
774145
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

97%

bp

214-216 °C/12 mmHg (lit.)

mp

25-30 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCBr

InChI

1S/C18H37Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

WSULSMOGMLRGKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Shahruzzaman et al.
Journal of separation science, 38(14), 2403-2413 (2015-05-07)
The amphiphilic polymer-grafted silica was newly prepared as a stationary phase in high-performance liquid chromatography. Poly(4-vinylpyridine) with a trimethoxysilyl group at one end was grafted onto porous silica particles and the pyridyl side chains were quaternized with 1-bromooctadecane. The obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service