Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900570

Sigma-Aldrich

2,3,5,6-Tetramethylpyrazine

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
Beilstein/REAXYS Number:
113100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

190 °C (lit.)

mp

77-80 °C (lit.)

SMILES string

Cc1nc(C)c(C)nc1C

InChI

1S/C8H12N2/c1-5-6(2)10-8(4)7(3)9-5/h1-4H3

InChI key

FINHMKGKINIASC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuesong Zhang et al.
Journal of cellular physiology (2019-02-17)
Inadequate oxygen supply is probably one of the most important pathophysiological mechanisms of cardiomyocyte damage in ischemic heart disease. Tetramethylpyrazine (TMP, also known as ligustrazine) is the main active ingredient isolated from the rhizome of Ligusticum chuanxiong Hort. A previous
Yu-Fang Leng et al.
The American journal of Chinese medicine, 40(6), 1229-1239 (2012-12-12)
The Bennett and Xie (1988) model of chronic constriction injury (CCI) investigated the effects of tetramethylpyrazine (TMP) on neuropathic pain-associated behaviors and neuronal apoptosis in the spinal dorsal horn. Fifty-four male rats were randomly divided into sham (group S), CCI
Jian-Zhong Hu et al.
Journal of the neurological sciences, 324(1-2), 94-99 (2012-11-13)
In the present study, we explored the effects of tetramethylpyrazine (TMP), an alkaloid extracted from the Chinese herbal medicine Ligusticum wallichii Franchat (chuanxiong), on a rat model of contusion spinal cord injury (SCI). The contusion SCI model was induced in
Yaogui Sun et al.
Pharmaceutical biology, 51(9), 1137-1143 (2013-04-24)
The prevalence of infectious bursal disease has brought about enormous financial losses to the world poultry industry. Chinese herb medicines can provide valuable materials for discovery and development of new drugs. To screen constituents derived from Chinese herb medicines for
Penglong Wang et al.
Die Pharmazie, 68(9), 782-789 (2013-10-24)
A new anticancer ligustrazine derivative, 3beta-hydroxyolea-12-en-28-oic acid-3,5,6-trimethylpyrazin-2-methyl ester (T-OA, C38H58O3N2), was previously reported. It was synthesized via conjugating the effective antitumor ingredients of a classic traditional Chinese medicine (TCM) formulation. In the present study, anticancer efficacy of T-OA was evaluated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service