Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900569

Sigma-Aldrich

10-Undecenoic acid

Vetec, reagent grade, 98%

Synonym(s):

Undecylenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
Colour Index Number:
42650
Beilstein/REAXYS Number:
1762631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.449 (lit.)

bp

137 °C/2 mmHg (lit.)

mp

23-25 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCCCCCCCC=C

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)

InChI key

FRPZMMHWLSIFAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

300.2 °F - closed cup

flash_point_c

149 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Guan et al.
Scientific reports, 5, 12641-12641 (2015-08-01)
This Report presents a nitrogen-doping method by chemically forming self-assembled monolayers on silicon. Van der Pauw technique, secondary-ion mass spectroscopy and low temperature Hall effect measurements are employed to characterize the nitrogen dopants. The experimental data show that the diffusion
Ying-Dan Hu et al.
Biotechnology progress, 30(5), 1005-1011 (2014-06-04)
Enzymatic synthesis of ascorbyl undecylenate, an unsaturated fatty acid ester of ascorbic acid, was reported with biomass-derived 2-methyltetrahydrofuran (MeTHF) as the cosolvent. Of the immobilized lipases tested, Candida antarctica lipase B (CAL-B) showed the highest activity for enzymatic synthesis of
Rongling Yang et al.
PloS one, 9(10), e110342-e110342 (2014-10-21)
Highly efficient and regioselective acylation of pharmacologically interesting gastrodin with vinyl undecylenic acid has been firstly performed through an enzymatic approach. The highest catalytic activity and regioselectivity towards the acylation of 7'-hydroxyl of gastrodin was obtained with Pseudomonas cepacia lipase.
Betar M Gallant et al.
ACS nano, 9(5), 5143-5153 (2015-04-16)
The interfacial shear strength between Si microwires and a Nafion membrane has been tailored through surface functionalization of the Si. Acidic (-COOH-terminated) or basic (-NH2-terminated) surface-bound functionality was introduced by hydrosilylation reactions to probe the interactions between the functionalized Si
S N Aisyiyah Jenie et al.
Biosensors & bioelectronics, 74, 637-643 (2015-07-24)
The up-regulation of L-lactate dehydrogenase (LDH), an intracellular enzyme present in most of all body tissues, is indicative of several pathological conditions and cellular death. Herein, we demonstrate LDH detection using porous silicon (pSi) microcavities as a luminescence-enhancing optical biosensing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service