Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900566

Sigma-Aldrich

Trimethyloctadecylammonium bromide

Vetec, reagent grade, 98%

Synonym(s):

Octadecyltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17N(Br)(CH3)3
CAS Number:
Molecular Weight:
392.50
Beilstein/REAXYS Number:
3574090
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

~250 °C (dec.) (lit.)

SMILES string

C[N+](C)(CCCCCCCCCCCCCCCCCC)C.[Br-]

InChI

1S/C21H46N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(2,3)4;/h5-21H2,1-4H3;1H/q+1;/p-1

InChI key

SZEMGTQCPRNXEG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan C López-Montilla et al.
Water research, 39(9), 1907-1913 (2005-05-19)
The removal of model pollutants bromocresol green (BG) and phenol from water is demonstrated via two liquid-liquid extraction methods. Both methods exploit selective interactions established by the pollutant molecule with a surfactant, oil, or alcohol, and are variants of the
Ken Nagai et al.
Colloids and surfaces. B, Biointerfaces, 56(1-2), 197-200 (2006-12-16)
We describe here our recent work on spontaneous regular motion of liquid droplet powered by the chemical Marangoni effect under spatially symmetric conditions. It is shown that a spontaneously crawling oil droplet on a glass substrate with a nonequilibrium chemical
Yutaka Sumino et al.
The journal of physical chemistry. B, 113(48), 15709-15714 (2009-11-12)
Spontaneous deformation of a tetradecane droplet with palmitic acid on an aqueous phase with stearyltrimethylammonium chloride is reported. Palmitic acid is transported from the oil droplet to the aqueous phase by the concentration difference between the organic and the aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service