Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900562

Sigma-Aldrich

N-Acetyl-DL-methionine

Vetec, reagent grade, 98%

Synonym(s):

Methionamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
191.25
Beilstein/REAXYS Number:
1867673
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

117-119 °C (lit.)

SMILES string

CSCCC(NC(C)=O)C(O)=O

InChI

1S/C7H13NO3S/c1-5(9)8-6(7(10)11)3-4-12-2/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)

Inchi Key

XUYPXLNMDZIRQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Kennepohl et al.
Journal of synchrotron radiation, 16(Pt 4), 484-488 (2009-06-19)
Despite a wealth of studies exemplifying the utility of the 2-5 keV X-ray range in speciation and electronic structure elucidation, the exploitation of this energy regime for the study of photochemical processes has not been forthcoming. Herein, a new endstation
D R Sheff et al.
The Journal of biological chemistry, 264(19), 11491-11496 (1989-07-05)
Genes for the various isoactins define two classes of actin. Class I actin genes code for Met-Asp(Glu)-actin, and class II actin genes code for Met-X-Asp(Glu)-actin where X is usually cysteine. Amino termini of both are removed in an acetylation-dependent processing
N-acetylmethionine and biotin as photocleavable protective groups for ruthenium polypyridyl complexes.
Roosmarijn E Goldbach et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(36), 9924-9929 (2011-07-29)
Adnan A Elfarra et al.
Biochimica et biophysica acta, 1703(2), 183-189 (2005-02-01)
Flavin-containing monooxygenases (FMOs) are microsomal enzymes that catalyze the NADPH-dependent oxidation of a large number of sulfur-, selenium-, and nitrogen-containing compounds. Five active isoforms (FMO1-5) have been identified and shown to be differently expressed in various mammalian tissues. Previous work
Rebecca D Sandlin et al.
Journal of inorganic biochemistry, 104(2), 214-216 (2009-11-13)
A bulky platinum triamine complex, [Pt(Me(5)dien)(NO(3))]NO(3) (Me(5)dien=N,N,N',N',N''-pentamethyldiethylenetriamine) has been prepared and reacted in D(2)O with N-acetylmethionine (N-AcMet) and guanosine 5'-monophosphate (5'-GMP); the reactions have been studied using (1)H NMR spectroscopy. Reaction with 5'-GMP leads to two rotamers of [Pt(Me(5)dien)(5'-GMP-N7)](+). Reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service