Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900561

Sigma-Aldrich

1,5-Pentanediol

Vetec, reagent grade, 96%

Synonym(s):

Pentamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)5OH
CAS Number:
Molecular Weight:
104.15
Beilstein/REAXYS Number:
1560130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor pressure

<0.01 mmHg ( 20 °C)

product line

Vetec

assay

96%

autoignition temp.

635 °F

expl. lim.

13.2 %

refractive index

n20/D 1.450 (lit.)

bp

242 °C (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

OCCCCCO

InChI

1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2

InChI key

ALQSHHUCVQOPAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

264.2 °F - closed cup

flash_point_c

129 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Yoshida et al.
Chemical communications (Cambridge, England), 48(55), 6981-6983 (2012-06-07)
Highly chemo- and regioselective acylation of 2-aminopentane-1,5-diol derivatives has been achieved by organocatalysis. An acyl group can be chemoselectively introduced onto the sterically hindered secondary hydroxy group in the presence of the primary one by virtue of the molecular recognition
Regina Busch et al.
Advances in therapy, 26(7), 719-727 (2009-08-04)
Most episodes of recurrent herpes labialis are self-limited and mild, but can be troublesome when they occur frequently with painful and unsightly lesions. Therefore, there has been much interest in developing agents that can suppress outbreaks in addition to being
Jan Faergemann et al.
Acta dermato-venereologica, 85(3), 203-205 (2005-07-26)
Multi-resistance to antibiotic therapy and to biocides is becoming increasingly common, which has led to mounting concern worldwide regarding the future use of traditional antimicrobials. Diols or glycols also have antimicrobial effects. Pentane-1,5-diol has low oral toxicity, is essentially non-irritating
Hanne Evenbratt et al.
Acta dermato-venereologica, 89(2), 126-129 (2009-03-28)
The aim of this study was to compare pentane-1,5-diol and propane-1,2-diol used as absorption enhancers for cutaneously administered terbinafine. Fresh human skin samples were placed in a continuous flow diffusion cell with a gel containing terbinafine on top of the
David M Hunsicker et al.
Macromolecular rapid communications, 33(3), 232-236 (2011-12-17)
The Milstein catalyst has proven to be highly effective for the conversion of alcohols to esters, as well as alcohols and amines to amides and polyamides. We have recently found that the catalyst's range can be extended to very efficient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service