Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900557

Sigma-Aldrich

2,5-Hexanedione

Vetec, reagent grade, 98%

Synonym(s):

Acetonylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14
Beilstein/REAXYS Number:
506525
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

0.43 mmHg ( 20 °C)

product line

Vetec

assay

98%

refractive index

n20/D 1.425 (lit.)

bp

191 °C (lit.)

mp

−6-−5 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CCC(C)=O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

target_organs

Nervous system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Ghelli et al.
PloS one, 4(11), e7922-e7922 (2009-11-26)
Leber's hereditary optic neuropathy (LHON) is a maternally inherited blinding disease due to mitochondrial DNA (mtDNA) point mutations in complex I subunit genes, whose incomplete penetrance has been attributed to both genetic and environmental factors. Indeed, the mtDNA background defined
Kim Boekelheide et al.
Annual review of pharmacology and toxicology, 43, 125-147 (2002-12-10)
Now in its third decade of mechanistic investigation, testicular injury caused by 2,5-hexanedione (2,5-HD) exposure is a well-studied model with a rich database. The development of this model reflects the larger changes that have moved biology from a branch of
Xin Cheng et al.
Neurotoxicology, 33(5), 1239-1247 (2012-07-31)
Worldwide, n-hexane is an organic solvent widely used in numerous industries such as chemical engineering, pharmaceutical and cosmetic industry. 2,5-Hexanedione (2,5-HD) is the main metabolite of n-hexane. It is now gradually recognized that chronic exposure to n-hexane could harm the
Hideki Yamasaki et al.
Toxicological sciences : an official journal of the Society of Toxicology, 117(2), 449-456 (2010-07-10)
In the testis, developing germ cells are dependent on supportive physical and paracrine interactions with Sertoli cells. The intimate nature of this relationship is demonstrated by the fact that a toxic insult compromising the stability of Sertoli cells will have
Min-Sun Kim et al.
Toxicology, 260(1-3), 97-103 (2009-05-26)
2,5-Hexanedione (HD), a metabolite of n-hexane, causes central and peripheral neuropathy leading to motor neuron deficits. Although chronic exposure to n-hexane is known to cause gradual sensorimotor neuropathy, there are no reports on the effects of low doses of HD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service