Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900556

Sigma-Aldrich

3-Hydroxypropionitrile

Vetec, reagent grade, 96%

Synonym(s):

2-Cyanoethanol, Ethylene cyanohydrin, Hydracrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CN
CAS Number:
Molecular Weight:
71.08
Beilstein/REAXYS Number:
635773
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

2.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

Vetec

assay

96%

autoignition temp.

922 °F

expl. lim.

12.1 %

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

mp

−46 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

OCCC#N

InChI

1S/C3H5NO/c4-2-1-3-5/h5H,1,3H2

InChI key

WSGYTJNNHPZFKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kid Törnquist et al.
European journal of pharmacology, 453(1), 1-11 (2002-10-24)
Sphingolipid derivatives cause diverse effects towards the regulation of intracellular free Ca(2+) concentrations ([Ca(2+)](i)) in a multitude of nonexcitable cells. In the present investigation, the effect of C-8 ceramide-1-(2-cyanoethyl) phosphate (C1CP) on store-operated Ca(2+) (SOC) entry was investigated. C1CP evoked
M Duverger-Van Bogaert et al.
Toxicology letters, 7(4-5), 311-319 (1981-02-01)
The mutagenicity of acrylonitrile (ACN) was tested with Salmonella typhimurium TA1530 after a preincubation period of the chemical with a rat liver post-mitochondrial fraction in liquid medium. Several pretreatments were applied to the animals before the preparation of the liver
M Lambotte-Vandepaer et al.
Toxicology letters, 7(4-5), 321-327 (1981-02-01)
Urines collected from rats injected with acrylonitrile (ACN) were mutagenic towards Salmonella typhimurium TA1530; this activity was reduced when the animals were pretreated by pyrazole (inhibitor of alcohol dehydrogenase) and suppressed after pretreatment either by CoCl2 and SKF 525-A (inhibitors
Ewa Radzikowska et al.
Organic & biomolecular chemistry, 13(1), 269-276 (2014-11-05)
Chimeric oligonucleotides containing phosphodiester and phosphorothioate linkages have been obtained using the solid phase synthesis. The oligonucleotide parts possessing natural internucleotide phosphate bonds were assembled using commercially available nucleoside 3'-O-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidites 7 whereas the phosphorothioate segment was built using nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholanes)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service