Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900554

Sigma-Aldrich

Glutaric anhydride

Vetec, reagent grade, 94%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
Beilstein/REAXYS Number:
110051
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

94%

bp

150 °C/10 mmHg (lit.)

mp

50-55 °C (lit.)

SMILES string

O=C1CCCC(=O)O1

InChI

1S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2

Inchi Key

VANNPISTIUFMLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Bowen et al.
Journal of dental research, 79(4), 905-911 (2000-06-01)
Polymerizable cyclodextrin derivatives (PCDs) have been proposed as candidates for use in dental therapeutics (Bowen, 1996; Bowen and Reed, 1997). Here, PCD "libraries" were synthesized by quasi-random reactions of 6 moles of methacrylic anhydride plus 6 moles of cyclic glutaric
W L Shelver et al.
Journal of immunoassay, 21(1), 1-23 (2000-05-11)
Antibody generated from ractopamine-hemiglutarate-KLH was used to develop a ractopamine ELISA. The antibody showed good sensitivity in phosphate buffer, with an IC50 of 4.2 ng/ml (ppb) toward ractopamine and 16.2 ng/ml toward glucuronides of ractopamine conjugated to the phenethanolamine phenol
Rüdiger Benters et al.
Nucleic acids research, 30(2), E10-E10 (2002-01-15)
The DNA microarray-based analysis of single nucleotide polymorphisms (SNPs) is important for the correlation of genetic variations and individual phenotypes, and for locating disease-causing genes. To facilitate the development of surfaces suitable for immobilization of oligonucleotides, we report here a
H B Lazrek et al.
Nucleosides, nucleotides & nucleic acids, 26(8-9), 1103-1106 (2007-12-07)
We report the synthesis of homo and heterodimers of 3TC conjugates. All new dimers were screened for their ability to inhibit HIV-1 in MT4 cell line and were compared to AZT alone and showed marked antiviral activity.
T Tanaka et al.
Biological & pharmaceutical bulletin, 16(12), 1270-1275 (1993-12-01)
The tumor distribution and the disposition of serum proteins, such as albumin, fetuin, transferrin, and IgG, were investigated in mice bearing Sarcoma 180. Serum proteins labeled with fluoresceinisothiocyanate (FITC) were administered to the mice. The FITC-labeled proteins acylated with glutaric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service