Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900553

Sigma-Aldrich

1,3-Dibromobenzene

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Br2
CAS Number:
Molecular Weight:
235.90
Beilstein/REAXYS Number:
1904538
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

8.16 (vs air)

vapor pressure

5 mmHg ( 66 °C)

product line

Vetec

assay

97%

refractive index

n20/D 1.608 (lit.)

bp

218-219 °C (lit.)

mp

−7 °C (lit.)

density

1.952 g/mL at 25 °C (lit.)

SMILES string

Brc1cccc(Br)c1

InChI

1S/C6H4Br2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

JSRLURSZEMLAFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

200.1 °F - closed cup

flash_point_c

93.40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sapota et al.
Chemosphere, 39(13), 2229-2238 (1999-11-27)
The distribution, excretion and metabolism of 1,3-dibromobenzene following a single i.p. administration to rats 100 or 300 mg/kg was investigated using radiotracer [3H] and GC-MS technique. After 72 hours about 74 to 90% were excreted in urine. The highest radioactivity
J A Szymańska
Archives of toxicology, 71(1-2), 99-106 (1996-01-01)
Rats were used to study acute and subacute hepatotoxicity of 1,3-dibromobenzene (1,3-dBB). In the single-exposure experiment, maximum hepatic 1,3-dBB concentrations were found to occur 1 to 12 h after the exposure, depending on the dose. Maximum concentrations of covalently bound
J A Szymańska et al.
Journal of applied toxicology : JAT, 16(1), 35-41 (1996-01-01)
Various doses of dibromobenzene isomers (1,2-dBB, 1,3-dBB, 1,4-dBB) were administered (i.p.) to BALB mice. The levels of reduced glutathione (GSH) and malondialdehyde (MDA) in the liver, and glutamate-pyruvate transaminase (GPT) (EC.2.6.1.2) gamma-glutamyltransferase (gamma-GT) (EC.2.3.2.2) and triglycerides (TG) in the serum
J A Szymańska
Archives of toxicology, 72(2), 97-103 (1998-02-10)
Brominated benzenes appear in the environment and human tissues. Their detection in the environment may be as a result of their usage, e.g. hexabromobenzene (HBB), and as products of HBB degradation or metabolism. The aim of this study was to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service