Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900550

Sigma-Aldrich

4-Hydroxycoumarin

Vetec, reagent grade, 98%

Synonym(s):

4-Hydroxy-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Beilstein/REAXYS Number:
129768
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

211-213 °C (lit.)

SMILES string

OC1=CC(=O)Oc2ccccc12

InChI

1S/C9H6O3/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5,10H

InChI key

VXIXUWQIVKSKSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen A Nolan et al.
Journal of medicinal chemistry, 52(22), 7142-7156 (2009-11-03)
The synthesis is reported here of two novel series of inhibitors of human NAD(P)H quinone oxidoreductase-1 (NQO1), an enzyme overexpressed in several types of tumor cell. The first series comprises substituted symmetric dicoumarol analogues; the second series contains hybrid compounds
Davorka Zavrsnik et al.
Bosnian journal of basic medical sciences, 8(3), 277-281 (2008-09-26)
Due to exceptional reactivity of 4-hydroxycoumarin, the synthesis of new coumarin derivatives of dimer and tetramer type has been carried out. The synthesis was carried out from 4-hydroxycoumarin and various aromatic aldehydes. In this way, compounds of the dimer 3,3'-(benzilidene)bis
Ana Carolina Luchini et al.
Biological & pharmaceutical bulletin, 31(7), 1343-1350 (2008-07-02)
Coumarins represent an important class of phenolic compounds with multiple biological activities, including inhibition of lipidic peroxidation and neutrophil-dependent anion superoxide generation, anti-inflammatory and immunosuppressor actions. All of these proprieties are essential for that a drug may be used in
Dharmesh H Mahajan et al.
Archiv der Pharmazie, 342(5), 281-290 (2009-05-06)
Novel 2-(coumarin-4-yloxy)-4,6-(substituted)-s-triazine derivatives i. e., diaryltriazine (DATA) are reported as novel non-nucleoside reverse transcriptase inhibitors (NNRTIs), were synthesized and their activities against human immunodeficiency virus HIV-1 (III-B), HIV-2 (ROD), and the double RT mutant HIV-1 (K103N and Y181C) were assessed.
Katrin J Czogalla et al.
Blood, 122(15), 2743-2750 (2013-08-29)
Since the discovery of warfarin-sensitive vitamin K 2,3-epoxide reductase complex subunit 1 (VKORC1), 26 human VKORC1 (hVKORC1) missense mutations have been associated with oral anticoagulant resistance (OACR). Assessment of warfarin resistance using the "classical" dithiothreitol-driven vitamin K 2,3-epoxide reductase (VKOR)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service