Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900547

Sigma-Aldrich

Dimethyl succinate

Vetec, reagent grade, 98%

Synonym(s):

Succinic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH2COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
956776
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

0.3 mmHg ( 20 °C)

product line

Vetec

assay

98%

autoignition temp.

689 °F

expl. lim.

8.5 %

refractive index

n20/D 1.419 (lit.)

bp

200 °C (lit.)

mp

16-19 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Leclercq-Meyer et al.
International journal of molecular medicine, 6(2), 143-152 (2000-07-13)
D-mannoheptulose was recently proposed to be transported into cells by GLUT2, whereas its hexaacetate ester may cross the plasma membrane without requiring the intervention of a specific carrier system. In the light of these proposals, the effects of unesterified D-mannoheptulose
Dushyant Shekhawat et al.
Bioresource technology, 97(2), 342-347 (2005-09-21)
A complete process model and economic analysis has been developed for itaconic acid production via catalytic condensation of dimethyl succinate and formaldehyde. The process model is based on experimental yields and selectivities obtained for the condensation reaction and on recovery
W J Malaisse et al.
Nutrition (Burbank, Los Angeles County, Calif.), 13(4), 330-341 (1997-04-01)
In rats injected with bacterial lipopolysaccharide (LPS; 5 gamma mg/g body weight [BWT]), the toxin provokes death within 24 h in 23% of the animals and, in surviving rats, causes a decrease in BWT, hyperlactacidemia, hyperlipacidemia, and hyperketonemia, as well
Michael J MacDonald
American journal of physiology. Endocrinology and metabolism, 283(2), E302-E310 (2002-07-12)
Succinic acid methyl esters are potent insulin secretagogues in rat pancreatic islets, but they do not stimulate insulin release in mouse islets. Unlike rat and human islets, mouse islets lack malic enzyme and, therefore, are unable to form pyruvate from
V Leclercq-Meyer et al.
Biochemical and molecular medicine, 59(1), 87-90 (1996-10-01)
Glucagon-like peptide 1 (GLP-1) is often referred to as a glucose-dependent insulinotropic agent and is currently under investigation as a tool in the treatment of noninsulin-dependent diabetes. This report shows that, in the absence of glucose, a nonglucidic nutrient, namely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service